Assay Details
| Gene Symbol: Msl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107048.1 | NP_001100518.1 | 1 - 2 | 805 | 67 | |
| XM_006247304.2 | XP_006247366.1 | 1 - 2 | 1124 | 67 | ||
| GenBank mRNA | FQ228290.1 | - | 1 - 2 | 759 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Msl1
Genomic Map