Assay Details
| Gene Symbol: Sap30bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108305.1 | NP_001101775.1 | 1 - 2 | 138 | 61 | |
| GenBank mRNA | BC169106.1 | - | 107 | 7458371 | 61 | |
| FQ220147.1 | - | 1 - 2 | 135 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sap30bp
Genomic Map