Assay Details
| Gene Symbol: Syngr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053553.1 | NP_446005.1 | 2 - 3 | 515 | 66 | |
| GenBank mRNA | AF039085.1 | - | 2 - 3 | 515 | 66 | |
| FQ218897.1 | - | 358 | 66 | |||
| FQ224907.1 | - | 359 | 66 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Syngr2
Genomic Map