Assay Details
| Gene Symbol: Nme1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138548.2 | NP_612557.1 | 1 - 2 | 99 | 66 | |
| GenBank mRNA | D13374.1 | - | 1 - 2 | 100 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the D13374.1, NM_138548.2 transcripts.
Set Membership: |
|

Nme1
Genomic Map