Assay Details
| Gene Symbol: Cacng4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080692.1 | NP_542423.1 | 1 - 2 | 216 | 77 | |
| GenBank mRNA | AF361341.1 | - | 1 - 2 | 216 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cacng4
Genomic Map