Assay Details
| Gene Symbol: Cacng4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080692.1 | NP_542423.1 | 3 - 4 | 446 | 68 | |
| GenBank mRNA | AF361341.1 | - | 3 - 4 | 446 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P149276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603769 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182839 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164877 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876801 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309029 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81895 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65340 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142328 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P149277 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P149277 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P149277 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P13078 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 43990 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 43539 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 53972 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58106 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 77394 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 335 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10748463 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748462 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748461 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748460 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10748459 | - | - |
| Rat Genome 230 2.0 Array | 1370755_at | - | - |
| Rat Genome 230 2.0 Array | 1392275_at | - | - |
| Rat Genome 230 2.0 Array | 1390548_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1370755_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1390548_at | - | - |
| Rat Genome 230 2.0 Array | 1392275_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370755_at | - | - |
| Affymetrix HT Rat Focus Array | 1370755_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000036319.rn.1 | TC1000003996.rn.1 | RTA TC |
| Rat Expression Set 230 | 1370755_PM_at | - | - |
| Rat Expression Set 230 | 1370755_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748459 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21598063 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1649760 | - | - |
| RatRef-12 Expression BeadChip | ILMN_62905 | - | - |

Cacng4
Genomic Map