Assay Details
| Gene Symbol: Esd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106051.3 | NP_001099521.1 | 2 - 3 | 125 | 154 | |
| NM_001270866.1 | NP_001257795.1 | 2 - 3 | 125 | 154 | ||
| GenBank mRNA | BC158790.1 | - | 2 - 3 | 116 | 7308742 | 154 |
| FQ213565.1 | - | 2 - 3 | 125 | 154 | ||
| FQ229329.1 | - | 2 - 3 | 99 | 154 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC158790.1,NM_001106051.3,NM_001270866.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Esd
Genomic Map