Assay Details
| Gene Symbol: Rhoq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053522.1 | NP_445974.1 | 4 - 5 | 769 | 63 | |
| GenBank mRNA | AB031482.1 | - | 4 - 5 | 769 | 63 | |
| BC061760.1 | - | 4 - 5 | 759 | 5621223 | 63 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
metabolic process
insulin receptor signaling pathway
regulation of cell shape
cortical actin cytoskeleton organization
cellular response to insulin stimulus
positive regulation of transcription from RNA polymerase II promoter
positive regulation of glucose import
positive regulation of filopodium assembly
negative regulation of establishment of protein localization to plasma membrane
GTPase activity
profilin binding
GTP binding
GBD domain binding
small GTPase mediated signal transduction
metabolic process
insulin receptor signaling pathway
regulation of cell shape
cortical actin cytoskeleton organization
cellular response to insulin stimulus
positive regulation of transcription from RNA polymerase II promoter
positive regulation of glucose import
positive regulation of filopodium assembly
negative regulation of establishment of protein localization to plasma membrane
GTPase activity
profilin binding
GTP binding
GBD domain binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P344475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172182 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330319 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499042 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29845 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P344475 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P344475 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P344475 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P344475 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12835 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700915503 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700416446 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55357 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 47723 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 2696 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49867 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700416446 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700915503 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10882753 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707387 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882754 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10888292 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10707386 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10882752 | - | - |
| Rat Genome 230 2.0 Array | 1367874_at | - | - |
| Rat Genome 230 2.0 Array | 1386967_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1367874_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1386967_at | - | - |
| Affymetrix HT Rat Focus Array | 1386967_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1367874_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1367874_at | - | - |
| Affymetrix HT Rat Focus Array | 1386967_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0600000868.rn.1 | TC0600000149.rn.1 | RTA TC |
| Rat Expression Set 230 | 1367874_PM_at | - | - |
| Rat Expression Set 230 | 1386967_PM_at | - | - |
| Rat Expression Set 230 | 1386967_at | - | - |
| Rat Expression Set 230 | 1367874_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882752 | - | - |
| Rat Genome U34 Set | rc_T26357_r_at | - | - |
| Rat Genome U34 Set | rc_T26357_i_at | - | - |
| Rat Genome U34 Set | rc_AA849961_at | - | - |
| Rat Genome U34 Set | T26358_at | - | - |
| Rat Genome U34 Set | rc_AI171999_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22181055 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357763 | - | - |
| RatRef-12 Expression BeadChip | ILMN_64423 | - | - |

Rhoq
Genomic Map