Assay Details
| Gene Symbol: Gpr113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106712.1 | NP_001100182.1 | 3 - 4 | 566 | 90 | |
| XM_006239800.2 | XP_006239862.1 | 4 - 5 | 839 | 90 | ||
| XM_006239801.2 | XP_006239863.1 | 3 - 4 | 698 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpr113
Genomic Map