Assay Details
| Gene Symbol: Gpr113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106712.1 | NP_001100182.1 | 7 - 8 | 1339 | 93 | |
| XM_006239800.2 | XP_006239862.1 | 8 - 9 | 1612 | 93 | ||
| XM_006239801.2 | XP_006239863.1 | 7 - 8 | 1471 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpr113
Genomic Map