Assay Details
| Gene Symbol: Fez2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053600.2 | NP_446052.2 | 2 - 3 | 380 | 87 | |
| XM_006239637.2 | XP_006239699.1 | 2 - 3 | 431 | 87 | ||
| XM_006239638.2 | XP_006239700.1 | 2 - 3 | 431 | 87 | ||
| XM_006239639.2 | XP_006239701.1 | 2 - 3 | 431 | 87 | ||
| GenBank mRNA | AB076183.1 | - | 2 - 3 | 87 | ||
| AB080086.1 | - | 2 - 3 | 87 | |||
| BC100060.1 | - | 2 - 3 | 361 | 7128839 | 87 | |
| U64689.1 | - | 2 - 3 | 87 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Fez2
Genomic Map