Assay Details
| Gene Symbol: Arhgap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008764656.1 | XP_008762878.1 | 2 - 3 | 3883 | 133 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgap5| Gene Symbol: Arhgap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008764656.1 | XP_008762878.1 | 2 - 3 | 3883 | 133 | |
Set Membership: |
|