Assay Details
| Gene Symbol: Slc7a15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106714.1 | NP_001100184.1 | 4 - 5 | 1172 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc7a15| Gene Symbol: Slc7a15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106714.1 | NP_001100184.1 | 4 - 5 | 1172 | 69 | |
Set Membership: |
|