Assay Details
| Gene Symbol: Asxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008764542.1 | XP_008762764.1 | 9 - 10 | 1517 | 66 | |
| XM_008764543.1 | XP_008762765.1 | 8 - 9 | 1373 | 66 | ||
More Information
Set Membership: |
|

Asxl2
Genomic Map