Assay Details
| Gene Symbol: Mgat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053604.2 | NP_446056.1 | 1 | 2235 | 90 | |
| GenBank mRNA | BC081754.1 | - | 1 | 2235 | 7130336 | 90 |
| U21662.1 | - | 1 | 1898 | 90 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mgat2
Genomic Map