Assay Details
| Gene Symbol: Pfn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009503.1 | NP_001009503.1 | 1 - 2 | 1003 | 75 | |
| GenBank mRNA | AY682392.1 | - | 1 - 2 | 1003 | 75 | |
| BC107465.1 | - | 1 - 2 | 1135 | 7453892 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pfn4
Genomic Map