Assay Details
| Gene Symbol: LOC103689957 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006239732.2 | XP_006239794.1 | 3 - 4 | 2135 | 82 | |
| XM_006239733.2 | XP_006239795.1 | 3 - 4 | 2132 | 82 | ||
| Gene Symbol: Fbxo11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181631.2 | NP_853662.1 | 3 - 4 | 351 | 82 | |
| XM_008764477.1 | XP_008762699.1 | 3 - 4 | 2125 | 82 | ||
| GenBank mRNA | AY274810.1 | - | 3 - 4 | 351 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC103689957
Genomic Map