Assay Details
| Gene Symbol: Slc26a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019214.1 | NP_062087.1 | 8 - 9 | 1167 | 77 | |
| XM_008764576.1 | XP_008762798.1 | 9 - 10 | 2491 | 77 | ||
| XM_008764577.1 | XP_008762799.1 | 7 - 8 | 1258 | 77 | ||
| GenBank mRNA | AF167412.1 | - | 8 - 9 | 1167 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc26a4
Genomic Map