Assay Details
| Gene Symbol: Fam150b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109372.1 | NP_001102842.1 | 2 - 3 | 749 | 140 | |
| GenBank mRNA | BC167018.1 | - | 2 - 3 | 790 | 7307720 | 140 |
More Information
Updated Mapping Information
Set Membership: |
|

Fam150b
Genomic Map