Assay Details
| Gene Symbol: Galm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007704.1 | NP_001007705.1 | 1 - 2 | 242 | 107 | |
| GenBank mRNA | BC081876.1 | - | 1 - 2 | 242 | 7107751 | 107 |
More Information
Updated Mapping Information
Set Membership: |
|

Galm
Genomic Map