Assay Details
| Gene Symbol: Prkd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024263.2 | NP_001019434.2 | 4 - 5 | 1315 | 72 | |
| XM_006239628.2 | XP_006239690.1 | 5 - 6 | 1476 | 72 | ||
| XM_006239629.2 | XP_006239691.1 | 5 - 6 | 1581 | 72 | ||
| XM_006239631.2 | XP_006239693.1 | 4 - 5 | 1432 | 72 | ||
| XM_008764421.1 | XP_008762643.1 | 5 - 6 | 1360 | 72 | ||
| GenBank mRNA | BC092663.1 | - | 3 - 4 | 520 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prkd3
Genomic Map