Assay Details
| Gene Symbol: Psma6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017283.3 | NP_058979.1 | 2 - 3 | 228 | 64 | |
| XM_006240098.2 | XP_006240160.1 | 2 - 3 | 186 | 64 | ||
| GenBank mRNA | BC062232.1 | - | 2 - 3 | 229 | 6888689 | 64 |
| D10755.1 | - | 2 - 3 | 217 | 64 | ||
| FQ212808.1 | - | 330 | 64 | |||
| FQ217234.1 | - | 2 - 3 | 204 | 64 | ||
| FQ220193.1 | - | 2 - 3 | 255 | 64 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Psma6
Genomic Map