Assay Details
| Gene Symbol: Ankmy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108019.1 | NP_001101489.1 | 3 - 4 | 460 | 73 | |
| XM_006240042.2 | XP_006240104.1 | 3 - 4 | 2845 | 73 | ||
| XM_006240043.2 | XP_006240105.2 | 4 - 5 | 3162 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ankmy2
Genomic Map