Assay Details
| Gene Symbol: Fkbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106736.2 | NP_001100206.2 | 1 - 2 | 129 | 131 | |
| GenBank mRNA | BC168873.1 | - | 1 - 2 | 94 | 131 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fkbp3
Genomic Map