Assay Details
| Gene Symbol: Fkbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106736.2 | NP_001100206.2 | 5 - 6 | 543 | 156 | |
| GenBank mRNA | BC168873.1 | - | 5 - 6 | 508 | 156 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fkbp3
Genomic Map