Assay Details
| Gene Symbol: Galnt14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012109.1 | NP_001012109.1 | 14 - 15 | 1709 | 87 | |
| XM_006239775.2 | XP_006239837.1 | 14 - 15 | 1876 | 87 | ||
| GenBank mRNA | BC079128.1 | - | 14 - 15 | 1709 | 7127183 | 87 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P091573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230876 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108042 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158520 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447996 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730838 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396511 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56177 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831116 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949569 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273351 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378235 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688809 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853079 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169901 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735554 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637802 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724842 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885877 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501497 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699733 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845497 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320489 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811410 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330872 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890029 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600326 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64785 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150655 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117352 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71213 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P091573 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P091573 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P091573 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 37879 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63884 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 104708 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55463 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63381 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 29091 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 45811 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 34335 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 70538 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 90744 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 74880 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 21416 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 73050 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38167 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 72788 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10882992 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882983 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882981 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882986 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882988 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882987 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882994 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882985 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882984 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882993 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882995 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882990 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882989 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882979 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882982 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882980 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10882978 | - | - |
| Rat Genome 230 2.0 Array | 1384676_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1396238_at | - | - |
| Rat Genome 230 2.0 Array | 1384676_at | - | - |
| Rat Genome 230 2.0 Array | 1396238_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1396238_at | - | - |
| Affymetrix HT Rat Focus Array | 1396238_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0600014203.rn.1 | TC0600001988.rn.1 | RTA TC |
| Rat Expression Set 230 | 1396238_PM_at | - | - |
| Rat Expression Set 230 | 1396238_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882978 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20780822 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376873 | - | - |

Galnt14
Genomic Map