Assay Details
| Gene Symbol: Bre | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199270.1 | NP_954891.1 | 3 - 4 | 292 | 63 | |
| XM_006239802.2 | XP_006239864.1 | 4 - 5 | 401 | 63 | ||
| GenBank mRNA | BC061573.1 | - | 3 - 4 | 292 | 6887072 | 63 |
More Information
Updated Mapping Information
Set Membership: |
|

Bre
Genomic Map