Assay Details
| Gene Symbol: Cript | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019907.1 | NP_063972.1 | 2 - 3 | 141 | 93 | |
| GenBank mRNA | AF047384.1 | - | 2 - 3 | 141 | 93 | |
| FQ222669.1 | - | 2 - 3 | 174 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cript
Genomic Map