Assay Details
| Gene Symbol: Actn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170325.1 | NP_001163796.1 | 4 - 5 | 640 | 87 | |
| GenBank mRNA | CK355337.1 | - | 4 - 5 | 640 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Actn2
Genomic Map