Assay Details
| Gene Symbol: Rdm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105842.1 | NP_001099312.1 | 3 - 4 | 431 | 74 | |
| GenBank mRNA | FQ215272.1 | - | 3 - 4 | 364 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rdm1
Genomic Map