Assay Details
| Gene Symbol: Lsm11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173384.1 | NP_001166855.1 | 3 - 4 | 689 | 154 | |
| GenBank mRNA | BC158836.1 | - | 3 - 4 | 689 | 7385359 | 154 |
| FQ223124.1 | - | 3 - 4 | 725 | 154 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lsm11
Genomic Map