Assay Details
| Gene Symbol: Med31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135813.1 | NP_001129285.1 | 3 - 4 | 264 | 143 | |
| GenBank mRNA | FQ211088.1 | - | 3 - 4 | 261 | 143 | |
More Information
Updated Mapping Information
Set Membership: |
|

Med31
Genomic Map