Assay Details
| Gene Symbol: Slc22a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019269.1 | NP_062142.1 | 7 - 8 | 1362 | 64 | |
| GenBank mRNA | AB017260.1 | - | 7 - 8 | 1362 | 64 | |
| AF110416.1 | - | 7 - 8 | 1362 | 64 | ||
| AJ001933.1 | - | 7 - 8 | 1348 | 64 | ||
| BC166421.1 | - | 6 - 7 | 1111 | 9089728 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a5
Genomic Map