Assay Details
| Gene Symbol: Ccl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012357.1 | NP_001012357.1 | 1 - 2 | 179 | 89 | |
| GenBank mRNA | AY863011.1 | - | 1 - 2 | 179 | 89 | |
| BC089865.1 | - | 1 - 2 | 179 | 7321336 | 89 | |
| EF121990.1 | - | 1 - 2 | 71 | 89 | ||
| EF121991.1 | - | 1 - 2 | 71 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccl9
Genomic Map