Assay Details
| Gene Symbol: Dcxr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134387.1 | NP_599214.1 | 3 - 4 | 321 | 82 | |
| XM_008768465.1 | XP_008766687.1 | 3 - 4 | 367 | 82 | ||
| XM_008768466.1 | XP_008766688.1 | 3 - 4 | 367 | 82 | ||
| XM_008768467.1 | XP_008766689.1 | 3 - 4 | 367 | 82 | ||
| GenBank mRNA | AB061719.1 | - | 3 - 4 | 321 | 82 | |
| FQ233401.1 | - | 3 - 4 | 320 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dcxr
Genomic Map