Assay Details
| Gene Symbol: Ttc19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109644.1 | NP_001103114.1 | 2 - 3 | 316 | 71 | |
| XM_006246530.2 | XP_006246592.1 | 2 - 3 | 294 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttc19
Genomic Map