Assay Details
Gene Symbol: LOC102554765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768453.1 | XP_008766675.1 | 3 - 4 | 917 | 66 |
Gene Symbol: Grin2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012575.3 | NP_036707.3 | 1 - 2 | 223 | 66 | |
XM_006247707.1 | XP_006247769.1 | 2 - 3 | 384 | 66 | ||
GenBank mRNA | D13212.1 | - | 1 - 2 | 223 | 66 | |
U08259.1 | - | 2 - 3 | 383 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the D13212.1, NM_012575.3, U08259.1, XM_006247707.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
