Assay Details
| Gene Symbol: Smim5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277479.1 | NP_001264408.1 | 2 - 3 | 328 | 72 | |
| GenBank mRNA | CB736773.1 | - | 2 - 3 | 328 | 72 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001277479.1 transcripts.
Set Membership: |
|

Smim5
Genomic Map