Assay Details
| Gene Symbol: Kpna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270802.1 | NP_001257731.1 | 4 - 5 | 434 | 123 | |
| NM_053483.2 | NP_445935.2 | 4 - 5 | 409 | 123 | ||
| GenBank mRNA | AJ130946.1 | - | 3 - 4 | 313 | 123 | |
| AY779026.1 | - | 3 - 4 | 303 | 123 | ||
| BC062026.1 | - | 4 - 5 | 409 | 5621957 | 123 | |
| BC089787.1 | - | 4 - 5 | 360 | 7326072 | 123 | |
| FM083931.1 | - | 4 - 5 | 409 | 123 | ||
| FM115539.1 | - | 4 - 5 | 431 | 123 | ||
| Gene Symbol: LOC100359600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729596.4 | XP_002729642.1 | 1 | 429 | 123 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Kpna2
Genomic Map