Assay Details
| Gene Symbol: Armcx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006257237.2 | XP_006257299.1 | 1 - 2 | 219 | 103 | |
| XM_006257238.2 | XP_006257300.1 | 1 - 2 | 219 | 103 | ||
| XM_006257240.2 | XP_006257302.1 | 1 - 2 | 220 | 103 | ||
| XM_006257241.2 | XP_006257303.1 | 1 - 2 | 219 | 103 | ||
| XM_008773455.1 | XP_008771677.1 | 1 - 2 | 219 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006257237.2, XM_006257238.2, XM_006257240.2, XM_006257241.2, XM_008773455.1 transcripts.
Set Membership: |
|

Armcx4
Genomic Map