Assay Details
| Gene Symbol: Armcx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006257237.2 | XP_006257299.1 | 4 - 5 | 453 | 127 | |
| XM_006257238.2 | XP_006257300.1 | 4 - 5 | 459 | 127 | ||
| XM_006257239.2 | XP_006257301.1 | 3 - 4 | 376 | 127 | ||
| XM_006257240.2 | XP_006257302.1 | 4 - 5 | 454 | 127 | ||
| XM_006257241.2 | XP_006257303.1 | 4 - 5 | 453 | 127 | ||
| XM_008773454.1 | XP_008771676.1 | 4 - 5 | 426 | 127 | ||
| XM_008773455.1 | XP_008771677.1 | 4 - 5 | 441 | 127 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006257237.2, XM_006257238.2, XM_006257239.2, XM_006257240.2, XM_006257241.2, XM_008773454.1, XM_008773455.1 transcripts.
Set Membership: |
|

Armcx4
Genomic Map