Assay Details
| Gene Symbol: Satl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163543.1 | NP_001157015.1 | 3 - 4 | 1960 | 94 | |
| XM_006257190.2 | XP_006257252.1 | 3 - 4 | 2407 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Satl1
Genomic Map