Assay Details
| Gene Symbol: LOC100910558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110680.1 | 1 - 2 | 278 | 68 | ||
| GenBank mRNA | FM089179.1 | - | 1 - 2 | 271 | 68 | |
| FM099320.1 | - | 1 - 2 | 229 | 68 | ||
| FQ215485.1 | - | 1 - 2 | 278 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

LOC100910558
Genomic Map