Assay Details
| Gene Symbol: Ttc39b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106665.1 | NP_001100135.1 | 16 - 17 | 1548 | 63 | |
| XM_006238350.2 | XP_006238412.1 | 15 - 16 | 2076 | 63 | ||
| XM_008763773.1 | XP_008761995.1 | 16 - 17 | 2199 | 63 | ||
| XM_008763774.1 | XP_008761996.1 | 17 - 18 | 1703 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttc39b
Genomic Map