Assay Details
| Gene Symbol: Pole3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007652.2 | NP_001007653.1 | 1 - 2 | 82 | 63 | |
| GenBank mRNA | BC081988.1 | - | 1 - 2 | 82 | 7129121 | 63 |
| BC083800.1 | - | 1 - 2 | 82 | 7108773 | 63 | |
| Gene Symbol: LOC100362333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729829.4 | XP_002729875.1 | 1 | 143 | 63 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pole3
Genomic Map