Assay Details
| Gene Symbol: Pigf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006239725.2 | XP_006239787.1 | 2 - 3 | 306 | 118 | |
| XM_008764472.1 | XP_008762694.1 | 2 - 3 | 345 | 118 | ||
More Information
Set Membership: |
|

Pigf
Genomic Map