Assay Details
| Gene Symbol: Pigf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006239725.2 | XP_006239787.1 | 3 - 4 | 395 | 147 | |
| XM_008764472.1 | XP_008762694.1 | 3 - 4 | 434 | 147 | ||
More Information
Set Membership: |
|

Pigf
Genomic Map