Assay Details
| Gene Symbol: Fkbp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022675.1 | NP_073166.1 | 1 - 2 | 39 | 68 | |
| GenBank mRNA | D86642.1 | - | 1 - 2 | 39 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fkbp1b
Genomic Map