Assay Details
| Gene Symbol: LOC103692570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008764540.1 | XP_008762762.1 | 1023 | 70 | ||
| Gene Symbol: Dpysl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023023.1 | NP_075412.1 | 7 - 8 | 912 | 70 | |
| GenBank mRNA | AB029432.1 | - | 6 - 7 | 786 | 70 | |
| AJ131436.1 | - | 7 - 8 | 912 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

LOC103692570
Genomic Map