Assay Details
| Gene Symbol: Fam179b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750170.3 | XP_003750218.1 | 18 - 19 | 5183 | 74 | |
| XM_006240139.2 | XP_006240201.1 | 17 - 18 | 5087 | 74 | ||
| XM_008764689.1 | XP_008762911.1 | 18 - 19 | 5237 | 74 | ||
| XM_234236.8 | XP_234236.4 | 19 - 20 | 5333 | 74 | ||
More Information
Set Membership: |
|

Fam179b
Genomic Map